Please login to your online account to display your discounted pricing

Human/Mouse Phospho-EphA5 DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC50372

 View more versions of this product

Catalog No. DYC50372

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 13839
Product Type ELISA Development Kit for phosphorylated intracellular proteins
Packaging Quantity 2 Plates
Quantity 1 Kit
Species Human, Mouse