missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human/Mouse Cleaved Caspase-3 (Asp175) DuoSet IC, R&D Systems

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

$416.79 - $822.18

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 12367
Species Human, Mouse
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
DYC8355 R & D Systems
RDSDYC8355
5 Plates Each for $822.18
DYC8352 R & D Systems
RDSDYC8352
2 Plates Each for $416.79
Specifications

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs See Product Certificates

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
1 Kit
Human, Mouse
ELISA Development Kit for intracellular proteins
12367