Please login to your online account to display your discounted pricing

eBiosciencetrade; Human MMP-3 Platinum ELISA Kit

For the quantitative detection of total human MMP-3.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Synonym Matrix metalloproteinase-3, Stromelysin-1, Progelatinase, Transin
Target MMP-3
Conjugate PE
Sample Type Serum, plasma (citrate, heparin), cell culture supernatant
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human MMP-3

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125245

 
ebioscience
LSBMS20143
96 tests Each for $651.05

501125246

 
ebioscience
LSBMS20143TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Matrix metalloproteinase-3, Stromelysin-1, Progelatinase, Transin
Target MMP-3
Conjugate PE
Sample Type Serum, plasma (citrate, heparin), cell culture supernatant
Sensitivity 0.0005ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.