Please login to your online account to display your discounted pricing

eBioscience™ Human MIP-1beta Instant ELISA Kit

For the quantitative detection of MIP-1beta levels in cell culture supernatants, serum and plasma.

$709.05 - $4,131.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym CCL4, LAG-1, Small inducible cytokine A4, SCYA4
Target MIP-1 beta
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with Polyclonal Antibody to MIP-1beta, Biotin-Conjugate (anti-MIP-1beta polyclonal antibody), Streptavidin-HRP and Sample diluent, lyophilized
Aluminium pouch(es) with a human MIP-1beta Standard curve (coloured)
Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Sample Diluent (Use when an external predilution of the samples is needed)
Stop Solution (1M Phosphoric acid)
Adhesive Plate Covers

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125259

 
life technologies
LSBMS2030INSTTEN
10 x 128 tests Each for $4,131.05

5017919

 
life technologies
LSBMS2030INST
128 tests Each for $709.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym CCL4, LAG-1, Small inducible cytokine A4, SCYA4
Target MIP-1 beta
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma (EDTA, heparin)
Sensitivity 3.0pg/mL
Species Human

Pre-coated plate(s)