Invitrogen™ eBioscience™ Human MIP-1beta Instant ELISA Kit
For the quantitative detection of MIP-1beta levels in cell culture supernatants, serum and plasma.
Manufacturer:  Invitrogen™ LSBMS2030INSTTEN
 View more versions of this product
Catalog No. 501125259
Includes: Aluminium pouch(es) with a Microwell Plate coated with Polyclonal Antibody to MIP-1beta, Biotin-Conjugate (anti-MIP-1beta polyclonal antibody), Streptavidin-HRP and Sample diluent, lyophilized
Aluminium pouch(es) with a human MIP-1beta Standard curve (coloured)
Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Sample Diluent (Use when an external predilution of the samples is needed)
Stop Solution (1M Phosphoric acid)
Adhesive Plate Covers
Description
Description
Pre-coated plate(s)
Specifications
Specifications
P13236
Biotin
6351
Pre-coated 96 well Microplate, Lysis Buffer 10x, Adhesive Plate Covers, Biotinylated Detection Antibody, SAV-HRP, Sample Diluent, lyophilized, Standard curve strips, Wash Buffer Concentrate, Substrate Solution, Sample Diluent, Stop Solution
10 x 128 tests
3.0pg/mL
50μL
Human
20 min
MIP-1 beta
CCL4, LAG-1, Small inducible cytokine A4, SCYA4
7.2%
7.8-500 pg/mL
ELISA
Colorimetric Microplate Reader
ELISA
RUO
Cell culture supernatant, serum, plasma (EDTA, heparin)
Plasma (50μL), Serum (50μL), Supernatants (50μL)
−20°C
3 hr 10 min
CCL4
4.9%
HRP