Please login to your online account to display your discounted pricing

eBiosciencetrade; Human MCP-1 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human MCP-1.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym Monocyte chemoattractant protein 1, MCAF, HC11
Target CCL2 (MCP-1)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human MCP-1
HRP-Conjugate anti-human MCP-1 monoclonal antibody
Human MCP-1 Standard lyophilized, 2ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125429

 
ebioscience
LSBMS281CE
96 tests Each for $665.55

501125431

 
ebioscience
LSBMS281TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym Monocyte chemoattractant protein 1, MCAF, HC11
Target CCL2 (MCP-1)
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, heparin)
Sensitivity 2.3pg/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.