Please login to your online account to display your discounted pricing

eBioscience™ Human M-CSF, Recombinant Protein, Carrier-Free

Human Recombinant Protein

$897.55 - $2,420.05

Specifications

Name M-CSF
Recombinant Yes
Conjugate Unlabeled
Purity >95%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123528

 
ebioscience™
LS34878982
100μg Each for $897.55

501123529

 
ebioscience™
LS34878985
500μg Each for $2,420.05
Description & Specifications

Specifications

Name M-CSF
Recombinant Yes
Conjugate Unlabeled
Purity >95%
Purification Method SDS-PAGE
Source Insect Cells
For Use With (Application) Cytokine Bioassays
Molecular Weight 36kDa
Storage Requirements Store at less than or equal to -70°C.

Human Macrophage Colony Stimulating Factor (M-CSF, also called CSF-1) is a ∽36 kDa disulfide-linked homodimer produced by a variety of cells including fibroblasts, macrophages, bone marrow stromal cells and endothelial cells. M-CSF regulates macrophage survival, proliferation and differentiation. M-CSF also enhances macrophage killing of tumor cells and microorganisms and regulates cytokine production from macrophages.