Please login to your online account to display your discounted pricing

Human M-CSF Quantikine ELISA Kit, R&D Systems

$663.51 - $3,180.69

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 1435
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Heparin Plasma (100μL), Saliva (100μL), Urine (100μL)
Sensitivity 47.3pg/mL
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity    

DMC00B

 
r & d systems
RDSDMC00B
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $663.51

SMC00B

 
r & d systems
RDSSMC00B
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $3,180.69
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 1435
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Heparin Plasma (100μL), Saliva (100μL), Urine (100μL)
Sensitivity 47.3pg/mL
Species Human

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Heparin Plasma (100μL), Saliva (100μL), Urine (100μL)
Sensitivity: 47.3pg/mL
Specificity: Natural and recombinant human M-CSF
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.