Please login to your online account to display your discounted pricing

eBioscience™ Human LAP (TGF beta 1) ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of LAP. TGF beta is synthesized as a precursor that contains LAP at the N-terminus and mature TGF beta at the C-terminus

$332.05 - $999.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Latency Associated Peptide, Pro-TGF beta 1, LAP/TGF beta 1
Target LAP
Sample Type Cell culture supernatant, serum and plasma (heparin)
View More Specs
Includes:  Capture Antibody Concentrate
Detection Antibody Concentrate
Human LAP standard (lyophilized): 20ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Sample Diluent B
pre-titrated Streptavidin-HRP
Tetramethylbenzidine (TMB) Substrate Solution

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

50246285

 
life technologies
LS885039022
96-well plates 2 x 96 tests Each for $332.05

50246286

 
life technologies
LS885039076
96-well plates 20 x 96 tests Each for $999.05

50246287

 
life technologies
LS885039077
Each for $941.05

50246289

 
life technologies
LS885039088
Each for $665.55

50246288

 
life technologies
LS885039086
96-well plates 10 x 96 tests Each for $694.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Latency Associated Peptide, Pro-TGF beta 1, LAP/TGF beta 1
Target LAP
Sample Type Cell culture supernatant, serum and plasma (heparin)
Sensitivity 156.3pg/mL
Storage Requirements 2° to 8°C
Species Human

LAP Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)