missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ING3 (aa 96-176) Control Fragment Recombinant Protein

Catalog No. RP103655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP103655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP103655 Supplier Invitrogen™ Supplier No. RP103655
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111622 (PA5-111622. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This gene can activate p53 trans-activated promoters, including promoters of p21/waf1 and bax. Overexpression of this gene has been shown to inhibit cell growth and induce apoptosis. Allelic loss and reduced expression of this gene were detected in head and neck cancers. Two alternatively spliced transcript variants encoding different isoforms have been observed.

Specifications

Accession Number Q9NXR8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54556
Name Human ING3 (aa 96-176) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1300013A07Rik; Eaf 4; Eaf4; FLJ20089; HSPC301; ING 2; ING 3; ING2; ING3; ing3 protein; inhibitor of growth family member 3; inhibitor of growth family, member 3; inhibitor of growth protein 3; inhibitor of growth protein 3-like protein; MEAF4; p47 p47 ING3; p47 regulator protein; p47ING3; p47ING3 protein; zgc:56327
Common Name ING3
Gene Symbol ING3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KLDQELAKFKMELEADNAGITEILERRSLELDTPSQPVNNHHAHSHTPVEKRKYNPTSHHTTTDHIPEKKFKSEALLSTLT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less