Please login to your online account to display your discounted pricing

eBiosciencetrade; Human IL-9 Platinum ELISA Kit

For the quantitative detection of human IL-9.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Interleukin 9, MEA, HP40, P40 cytokine
Target IL-9
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-9
Biotin-Conjugate anti-human IL-9 monoclonal antibody
Streptavidin-HRP
Human IL-9 Standard lyophilized, 200pg/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Sample Diluent
Calibrator Diluent
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5017985

 
ebioscience&trade
LSBMS2081TEN
10 x 96 tests Each for $4,189.05

5017984

 
ebioscience&trade
LSBMS2081
96 tests Each for $651.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Interleukin 9, MEA, HP40, P40 cytokine
Target IL-9
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 0.5pg/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.