Please login to your online account to display your discounted pricing

eBioscience Human IL-8 ELISA Ready-SET-Go!™ Kit, 2nd Generation

Specifically engineered for accurate and precise measurement of IL-8 protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity with a range from 2-250pg/mL.

$303.05 - $999.05

Specifications

For Use With (Application) ELISA
Synonym Interleukin-8, CXCL8, NAP-I
Target IL-8, IL-8 (CXCL8)
Sensitivity 2pg/mL
Storage Requirements 2° to 8°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and standards.

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

50246338

 
ebioscience
LS88808622
96-well plates 2 x 96 tests Each for $303.05

50246339

 
ebioscience
LS88808676
96-well plates 20 x 96 tests Each for $999.05

50246340

 
ebioscience
LS88808677
Each for $941.05

50246341

 
ebioscience
LS88808686
96-well plates 10 x 96 tests Each for $694.55

50246342

 
ebioscience
LS88808688
Each for $665.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Interleukin-8, CXCL8, NAP-I
Target IL-8, IL-8 (CXCL8)
Sensitivity 2pg/mL
Storage Requirements 2° to 8°C
Species Human

Human IL-8 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)