missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interleukin 8 Recombinant, Thermo Scientific™

Interleukin 8 Recombinant protein SIL8 from Thermo Scientific™ specifically detects Interleukin 8 in Not Applicable samples, and it is validated for Control, ELISA and WB

$651.05 - $2,436.00

Specifications

Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Sample Type Culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
Target Natural and recombinant human interleukin-8 (IL-8) and has been shown to recognize IL-8 derived from stimulated human umbilical vein endothelial cells
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Sufficient For Price Quantity  
Catalog Number Mfr. No. Description Sufficient For Price Quantity  
ENEH2IL8 Thermo Scientific™
PIEH2IL8
Human IL-8 ELISA Kit 96 microplate wells Each for $651.05
ENEH2IL82 Thermo Scientific™
EH2IL82
Human IL-8 ELISA Kit 2 x 96 microplate wells Each for $1,724.10
ENEH2IL85 Thermo Scientific™
PIEH2IL85
Human IL-8 ELISA Kit 5 x 96 microplate wells Pack of 5 Each for $2,436.00
Specifications

Description

Interleukin 8 Recombinant protein SIL8 from Thermo Scientific™ specifically detects Interleukin 8 in Not Applicable samples, and it is validated for Control, ELISA and WB

Chemokines are a group of small (approximately 8 to 14 kD), mostly basic, structurally related molecules that regulate cell trafficking of various types of leukocytes through interactions with a subset of 7-transmembrane, G protein-coupled receptors. Chemokines also play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. Chemokines are divided into 2 major subfamilies, CXC and CC, based on the arrangement of the first 2 of the 4 conserved cysteine residues; the 2 cysteines are separated by a single amino acid in CXC chemokines and are adjacent in CC chemokines. CCR10 is the receptor for CCL27 (SCYA27; MIM 604833); CCR10-CCL27 interactions are involved in T cell-mediated skin inflammation (Homey et al., 2002 [PubMed 11821900]).
Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 1 Vial purified Protein (2.5 ng/ml) in 4% BSA and no preservative;

PA5-32694 detects CCR10 in Human samples. PA5-32694 has been successfully used in ELISA and Immunohistochemistry (Paraffin) procedures. PA5-32694 The immunogen is Synthetic 17 amino acid peptide from C-terminus of human CCR10..
Description & Specifications

Specifications

Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Intra-assay CV <10%; inter-assay CV <10%
Natural and recombinant human interleukin-8 (IL-8) and has been shown to recognize IL-8 derived from stimulated human umbilical vein endothelial cells
Absorbance at 450nm minus absorbance at 550nm
Culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)