Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interleukin 8 Recombinant, Thermo Scientific™


Interleukin 8 Recombinant protein SIL8 from Thermo Scientific™ specifically detects Interleukin 8 in Not Applicable samples, and it is validated for Control, ELISA and WB

Manufacturer: thermo scientific™  PIEH2IL85

 View more versions of this product

Catalog No. ENEH2IL85

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Precision Intra-assay CV <10%; inter-assay CV <10%
Sensitivity <2pg/mL
Description Human IL-8 ELISA Kit
Assay Range 25.6 to 1000pg/mL
Quantity 5 plate kit
Target Natural and recombinant human interleukin-8 (IL-8) and has been shown to recognize IL-8 derived from stimulated human umbilical vein endothelial cells
Kit Contents 30X Wash Buffer, 200mL
Formulation Complete ELISA kit with coated 96-well plates
Measurement Absorbance at 450nm minus absorbance at 550nm
Sample Type Culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
Sufficient For 5 x 96 microplate wells
Specificity No significant cross-reaction with with human PF-4, GROa, IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-6, IL-7, IL-10, IL-12, TNFa, IFNa, IFNg, or GM-CSF
Sample Size 50µL
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Total Assay Time 3 hr.

Interleukin 8 Recombinant protein SIL8 from Thermo Scientific™ specifically detects Interleukin 8 in Not Applicable samples, and it is validated for Control, ELISA and WB

These human IL-8 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-8 (IL-8) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. The antibody-pair kit consists of the matched IL-8 antibody pairs and IL-8 standard only. All components undergo quality control testing to minimize lot-to-lot variability.

  • Target: Natural and recombinant human interleukin-8 (IL-8) and has been shown to recognize IL-8 derived from stimulated human umbilical vein endothelial cells

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 25.6 to 1000pg/mL

  • Sensitivity: < 2pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with with human PF-4, GROα, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-6, IL-7, IL-10, IL-12, TNFα, IFNα, IFNγ, or GM-CSF

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).