Please login to your online account to display your discounted pricing

Human IL-6 Quantikine HS ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSHS600B

 View more versions of this product

Catalog No. HS600B

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3569
Product Type Solid Phase Sandwich ELISA
Packaging Quantity 1 Plate
Quantity 1 Kit
Sensitivity 0.11pg/mL
Sample Type Serum, EDTA Plasma, Citrate Plasma, Urine
Sample Volume Serum (100μL), EDTA Plasma (100μL), Citrate Plasma (100μL), Urine (100μL)
Species Human
Specificity Natural and recombinant human IL-6
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.