missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interleukin 6 Recombinant, Thermo Scientific™

Interleukin 6 Recombinant protein SIL6 from Thermo Scientific™ specifically detects Interleukin 6 in Not Applicable samples, and it is validated for Control, ELISA and WB

Manufacturer:  Thermo Scientific™ SIL6

 View more versions of this product

Catalog No. ENSIL6


 

Specifications

Description

Interleukin 6 Recombinant protein SIL6 from Thermo Scientific™ specifically detects Interleukin 6 in Not Applicable samples, and it is validated for Control, ELISA and WB

Caveolae are specialized domains of the plasma membrane that are implicated in the sequestration of a variety of lipid and protein molecules. It has been suggested that these important cellular organelles have a pivotal role in such diverse biochemical processes as lipid metabolism, growth regulation, signal transduction, and apoptosis. Caveolin interacts with and regulates heterotrimeric G-proteins. Currently, there are three members of the caveolin multigene family which are known to encode 21-24 kDa integral membrane proteins that comprise the major structural component of the caveolar membrane in vivo. Caveolin-2 protein is abundantly expressed in fibroblasts and differentiated adipocytes, smooth and skeletal muscle, and endothelial cells. The expression of caveolin-1 is similar to that of caveolin-2 while caveolin-3 expression appears to be limited to muscle tissue types.
Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 1 Vial purified Protein (1 ng/ml) in 4% BSA and no preservative;

PA5-32691 detects CAV1/Caveolin in Human samples. PA5-32691 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-32691 The immunogen is Synthetic 16 amino acid peptide from N-terminus of human CAV1/Caveolin..
Description & Specifications

Specifications

<1 pg/mL
1Vial
store at -20°C, Avoid Freeze/Thaw Cycles
SIL6
50 µL
Absorbance at 450nm minus absorbance at 550nm
10.2 to 400 pg/mL
3 hrs
Intra-assay CV <10%; inter-assay CV <10%
1 Vial purified Protein (1 ng/ml) in 4% BSA and no preservative;
Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Natural and recombinant human interleukin-6 (IL-6)
ELISA standard curve with replicates
Human IL-6 Standard for ELISA