Please login to your online account to display your discounted pricing

eBioscience™ Human IL-4 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IL-4.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym B-cell stimulatory factor 1, B cell growth factor 1
Target IL-4
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-4
Biotin-Conjugate anti-human IL-4 monoclonal antibody
Streptavidin-HRP
Human IL-4 Standard lyophilized, 1ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125383

 
life technologies
LSBMS2252CE
96 tests Each for $665.55

501125384

 
life technologies
LSBMS2252TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym B-cell stimulatory factor 1, B cell growth factor 1
Target IL-4
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma, urine
Sensitivity 1.3pg/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.