missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ eBioscience™ Human IL-33 Platinum ELISA Kit
For the quantitative detection of human IL-33.
Manufacturer: Invitrogen™ LSBMS2048
Includes: Aluminium pouch(es) with a Microwell Plate coated with polyclonal polyclonal antibody to human IL-33
Biotin-Conjugate anti-human IL-33 polyclonal polyclonal antibody
Streptavidin-HRP
Human IL-33 Standard lyophilized, 500pg/mL upon reconstitution
Sample Diluent
Conjugate Diluent
Calibrator Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
O95760 | |
Biotin | |
90865 | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 tests | |
0.2pg/mL | |
50μL | |
Human | |
1 hr 20 min | |
IL-33 | |
Interleukin 33, DVS27-related protein, IL1F11 | |
4.7% |
7.8-500 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
ELISA | |
RUO | |
Cell culture supernatant, serum, plasma (citrate, heparin) | |
Plasma (50μL), Serum (50μL), Supernatants (50μL) | |
2-8°C | |
4 hr 30 min | |
IL33 | |
6.9% | |
HRP |
For Research Use Only