Please login to your online account to display your discounted pricing

Human IL-33 DuoSet, R&D Systems

Manufacturer: r & d systems  RDSDY3625

 View all options for this product

Catalog No. DY3625


Description & Specifications


Gene ID (Entrez) 90865

Assay Type: ELISA Development Kit
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.