Please login to your online account to display your discounted pricing

Human IL-33 DuoSet, R&D Systems

$846.01 - $846.01

Specifications

Gene ID (Entrez) 90865
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Quantity Price Quantity    

DY3625

 
r & d systems
RDSDY3625
Each for $846.01
Description & Specifications

Specifications

Gene ID (Entrez) 90865

Assay Type: ELISA Development Kit
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.