Please login to your online account to display your discounted pricing

eBioscience Human IL-29 (IFN lambda 1) ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of human IL-29 protein levels from samples including serum, plasma, and supernatants from cell cultures.

$332.05 - $941.05

Specifications

For Use With (Application) ELISA
Synonym Interleukin-29
Target IL-29 (IFN lambda)
Sensitivity 8pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISA Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20).
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

 Alert

Contains Azide

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

5017314

 
ebioscience
88729622
Each for $332.05

5017317

 
ebioscience
88729686
96-well plates 10 x 96 tests Each for $694.55

5017318

 
ebioscience
88729688
Each for $665.55

5017316

 
ebioscience
88729677
Each for $941.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Interleukin-29
Target IL-29 (IFN lambda)
Sensitivity 8pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Human

Human IL-29/IFN-\lambda Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)