Please login to your online account to display your discounted pricing

eBioscience Human IL-27 ELISA Ready-SET-Go!™ Kit

Allows the specific detection of IL-27 with no detection of IL-35 or free p28 or EBI3. This set has been optimized for the accurate and precise measurement of human IL-27 in serum, plasma, and tissue culture supernatant samples.

$332.05 - $694.55

Specifications

For Use With (Application) ELISA
Synonym Interleukin-27
Target IL-27
Sensitivity 64pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X PBS Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for the dilution of antibodies and standards

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

5017308

 
ebioscience
LS88727822
Each for $332.05

5017310

 
ebioscience
LS88727888
Each for $665.55

5017309

 
ebioscience
LS88727886
96-well plates 10 x 96 tests Each for $694.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Interleukin-27
Target IL-27
Sensitivity 64pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Human

Human IL-27 ELISA Ready-SET-Go!™ set includes all of the necessary buffers and reagents to perform enzyme-linked immunosorbent assays. Serum and plasma samples must be diluted 2-fold in prepared assay diluent prior to evaluation in this ELISA.