missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human IL-23 DuoSet, R&D Systems

contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant Human IL-23

$822.18 - $822.18

Specifications

Applications ELISA
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 kit
Species Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Format Price Quantity  
Catalog Number Mfr. No. Format Price Quantity  
DY1290 R & D Systems
RDSDY1290
15 96-well plates Each for $822.18
Specifications

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs See Product Certificates

Description & Specifications

Specifications

ELISA
ELISA kit
Human
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
1 kit