missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Human IL-22 Platinum ELISA Kit

For the quantitative detection of human IL-22.

$520.84 - $2,932.34

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym ILD110, ILTIF, TIFa
Target IL-22
Conjugate Biotin
View More Specs

Disclaimers:

For Research Use Only


Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-22
Biotin-Conjugate anti-human IL-22 monoclonal antibody
Streptavidin-HRP
Human IL-22 Standard lyophilized, 4ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5017954 Life Technologies
LSBMS2047
96 tests Each for $520.84
5017955 Life Technologies
LSBMS2047TEN
10 x 96 tests Each for $2,932.34
Specifications

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Description & Specifications

Specifications

ELISA
ILD110, ILTIF, TIFa
Biotin
5.0pg/mL
50μL
IL-22
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human