Please login to your online account to display your discounted pricing

eBioscience™ Human IL-22 ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of human IL-22 protein levels from samples including serum, plasma, and cell culture supernatants.

$332.05 - $590.37

Specifications

For Use With (Application) ELISA
Synonym Interleukin-22
Target IL-22
Sensitivity 8pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X concentrated.
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

501125209

 
life technologies
LS88752222
Each for $332.05

5017420

 
life technologies
LS88752286
96-well plates 10 x 96 tests Each for $590.37

5017421

 
life technologies
LS88752288
Each for $565.72
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Interleukin-22
Target IL-22
Sensitivity 8pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Human

Human IL-22 ELISA Ready-SET-Go! reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA)