missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Human IL-21 ELISA Ready-SET-Go!™ Kit, 2nd Generation

Specifically engineered for accurate and precise measurement of IL-21 protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity with a range from 8-1000pg/mL.

Manufacturer:  Invitrogen™ LS88821877

 View more versions of this product

Catalog No. 501125219


Alert:

Contains Sodium azide


Includes: Capture Antibody.Pre-titrated, purified antibody
Detection Antibody.Pre-titrated, biotin-conjugated antibody
Standard.Recombinant protein for generating standard curve and calibrating samples
Coating Buffer.10X PBS ELISA Coating Buffer
Assay Diluent.5X Concentrated
Detection Enzyme.Pre-titrated Avidin-HRP
Substrate Solution.Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and standards.

Description

Description

Human IL-21 Ready-SET-Go! (2nd Generation) ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

ELISA
8pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
Interleukin-21
20 x 96 tests
Human
IL-21
Documents

For Research Use Only