Please login to your online account to display your discounted pricing

eBiosciencetrade; Human IL-2 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IL-2.

$665.55 - $4,197.75

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Aldesleukin, T cell growth factor, TCGF Aldesleukin, T cell growth factor, TCGF
Target IL-2
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-2

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125364

 
ebioscience
LSBMS2212CE
96 tests Each for $665.55

501125367

 
ebioscience
LSBMS2212TENCE
10 x 96 tests Each for $4,197.75
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Aldesleukin, T cell growth factor, TCGF Aldesleukin, T cell growth factor, TCGF
Target IL-2
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), amniotic fluid, urine, whole blood
Sensitivity 9.2pg/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.