Please login to your online account to display your discounted pricing

Human IL-1ra/IL-1F3 DuoSet, R&D Systems

$324.58 - $846.01

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DY280

 
r & d systems
RDSDY280
15 Plates Each for $846.01

DY28005

 
r & d systems
RDSDY28005
5 Plates Each for $324.58
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit
Species Human

Assay Type: ELISA Development Kit
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.