Invitrogen™ eBioscience™ Human IL-17F (homodimer) ELISA Ready-SET-Go!™ Kit
Specifically engineered for accurate and precise measurement of human IL-17F homodimer protein levels from samples including serum, plasma and cell culture supernatants.
Manufacturer: Invitrogen™ LS88747886
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X Carbonate ELISA Coating Buffer
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution
Description
Human IL-17F ELISA Ready-SET-Go! Reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)- The assay demonstrates parallelism in measuring recombinant and native human IL-17F proteins and a standard curve range of 16 to 2000pg/mL
- Very specific to IL-17F homodimer. No crossreactivity to the IL-17A homodimer is observed and 0.24% crossreactivity to recombinant human IL-17AF heterodimer when spiked in excess (100ng/ml)
Specifications
ELISA | |
10 x 96 tests | |
Human | |
IL-17F |
96-well plates | |
16pg/mL | |
2° to 8°C except standard which should be stored at ≤−70°C | |
Interleukin-17F, IL-17FF |
For Research Use Only