missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interleukin 13 Recombinant, Thermo Scientific™

Interleukin 13 Recombinant protein SIL13 from Thermo Scientific™ specifically detects Interleukin 13 in samples, and it is validated for ELISA and Western Blot

$637.58 - $637.58

Specifications

Format Colorimetric ELISA (TMB substrate) in clear 96 well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Sample Type Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
Target Natural and recombinant human interleukin-13 (IL-13)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ENEHIL13 Thermo Scientific™
PIEHIL13
1 plate kit Each for $637.58
Specifications

Description

Interleukin 13 Recombinant protein SIL13 from Thermo Scientific™ specifically detects Interleukin 13 in samples, and it is validated for ELISA and Western Blot

Receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. This receptor stimulates chemotaxis, granule enzyme release and superoxide anion production.
Host Species: E. Coli
Formulation: 1 Vial purified Protein (2.5 ng/ml) in 4% BSA and no preservative;

PA5-32683 detects C5AR1/CD88 in Human samples. PA5-32683 has been successfully used in Immunohistochemistry and Immunohistochemistry (Paraffin) procedures. PA5-32683 The immunogen is Synthetic 18 amino acid peptide from internal region of human C5AR1/CD88..
Description & Specifications

Specifications

Colorimetric ELISA (TMB substrate) in clear 96 well strip plate
Intra-assay CV <10%; inter-assay CV <10%
Natural and recombinant human interleukin-13 (IL-13)
Absorbance at 450nm minus absorbance at 550nm
Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)