missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human IL-12 p70 Quantikine ELISA Kit, R&D Systems

a 3.5 or 4.5 hour solid phase ELISA designed to measure IL-12 in cell culture supernates, serum, plasma, and urine.

$644.82 - $3,091.10

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3592
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma, Urine
Sample Volume Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL), Urine (200μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
D1200 R & D Systems
RDSD1200
1 Plate Each for $644.82
S1200R R & D Systems
RDSS1200
6 Plates Each for $3,091.10
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL), Urine (200μL)
Sensitivity: 5pg/mL
Specificity: Natural and recombinant human IL-12p70
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
3592
Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL), Urine (200μL)
Human
Solid Phase Sandwich ELISA
Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma, Urine
5pg/mL
Natural and recombinant human IL-12p70