missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interleukin 12 (p70) Recombinant, Thermo Scientific™

Interleukin 12 (p70) Recombinant protein SIL12 from Thermo Scientific™ detects Interleukin 12 (p70) in Not Applicable samples, and is validated for Control, ELISA and WB

$175.45 - $2,436.00

Specifications

Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Sample Type Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ENEHIL125 Thermo Scientific™
PIEHIL125
5 plate kit Pack of 5 Each for $2,436.00
ENSIL12 Thermo Scientific™
SIL12
1Vial Each for $175.45
Specifications

Description

Interleukin 12 (p70) Recombinant protein SIL12 from Thermo Scientific™ detects Interleukin 12 (p70) in Not Applicable samples, and is validated for Control, ELISA and WB

CCR4 is a chemokine receptor and is preferentially expressed on type 2 helper T (Th2-type) cells. In contrast to other chemokine receptors, the expression of CCR4 and CCR8 on Th2 cells is transiently increased following TCR and CD28 engagement. Moreover, activated Th1 cells up-regulate CCR4 expression and functional responsiveness to thymus- and activation-regulated chemokine.
Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 1 Vial purified Protein (1.5 ng/ml) in 4% BSA and no preservative;

PA5-32697 detects CCR4 in Human samples. PA5-32697 has been successfully used in ELISA and Immunohistochemistry (Paraffin) procedures. PA5-32697 The immunogen is Synthetic 16 amino acid peptide from C-terminal cytoplasmic domain of human CCR4..
Description & Specifications

Specifications

Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Absorbance at 450nm minus absorbance at 550nm
Intra-assay CV <10%; inter-assay CV <10%
Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)