Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interleukin 12 (p70) Recombinant, Thermo Scientific™


Interleukin 12 (p70) Recombinant protein SIL12 from Thermo Scientific™ detects Interleukin 12 (p70) in Not Applicable samples, and is validated for Control, ELISA and WB

Manufacturer: thermo scientific™  SIL12

 View more versions of this product

Catalog No. ENSIL12

  • / Each
 

Description & Specifications

Specifications

Precision Intra-assay CV <10%; inter-assay CV <10%
Sensitivity <3 pg/mL
Formulation 1 Vial purified Protein (1.5 ng/ml) in 4% BSA and no preservative;
Quantity 1Vial
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Assay Range 15.4 to 600 pg/mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Sample Type Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
Sample Size 50 µL
Specificity SIL12
Sufficient For ELISA standard curve with replicates
Total Assay Time 3 hrs
Target SIL12
Description Human IL-12 (p70) Standard for ELISA

Interleukin 12 (p70) Recombinant protein SIL12 from Thermo Scientific™ detects Interleukin 12 (p70) in Not Applicable samples, and is validated for Control, ELISA and WB

CCR4 is a chemokine receptor and is preferentially expressed on type 2 helper T (Th2-type) cells. In contrast to other chemokine receptors, the expression of CCR4 and CCR8 on Th2 cells is transiently increased following TCR and CD28 engagement. Moreover, activated Th1 cells up-regulate CCR4 expression and functional responsiveness to thymus- and activation-regulated chemokine.
Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 1 Vial purified Protein (1.5 ng/ml) in 4% BSA and no preservative;

PA5-32697 detects CCR4 in Human samples. PA5-32697 has been successfully used in ELISA and Immunohistochemistry (Paraffin) procedures. PA5-32697 The immunogen is Synthetic 16 amino acid peptide from C-terminal cytoplasmic domain of human CCR4..