Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interleukin 12 (p70) Recombinant, Thermo Scientific™


Interleukin 12 (p70) Recombinant protein SIL12 from Thermo Scientific™ detects Interleukin 12 (p70) in Not Applicable samples, and is validated for Control, ELISA and WB

Manufacturer: thermo scientific™  PIEHIL125

 View more versions of this product

Catalog No. ENEHIL125

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Precision Intra-assay CV <10%; inter-assay CV <10%
Sensitivity <3pg/mL
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Formulation Complete ELISA kit with coated 96-well plates
Kit Contents 30X Wash Buffer, 200mL
Assay Range 15.4 to 600pg/mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Quantity 5 plate kit
Sample Type Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
Sample Size 50µL
Specificity No significant cross-reaction with human IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-13, IL-15, TNFa, IFNg, GM-CSF, GROa or GROß. Recombinant human IL-12 (p40) and mouse IL-12 (p40) do not cross-react or interfere with t
Sufficient For 5 x 96 microplate wells
Total Assay Time 3 hr.
Target Biologically active (p70) natural and recombinant human interleukin-12 (IL-12, p70)
Description Human IL-12 (p70) ELISA Kit

Interleukin 12 (p70) Recombinant protein SIL12 from Thermo Scientific™ detects Interleukin 12 (p70) in Not Applicable samples, and is validated for Control, ELISA and WB

These human IL-12 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of biologically active interleukin-12 (IL-12 p70) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

  • Target: Biologically active (p70) natural and recombinant human interleukin-12 (IL-12, p70)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 15.4 to 600pg/mL

  • Sensitivity: < 3pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-13, IL-15, TNFα, IFNγ, GM-CSF, GROα or GROβ. Recombinant human IL-12 (p40) and mouse IL-12 (p40) do not cross-react or interfere with this assay

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.