missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Human IL-10 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IL-10

$605.88 - $3,945.48

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym TGIF, MCSF III, CSIF, B TCGF
Target IL-10
Conjugate Biotin
View More Specs

Disclaimers:

For Research Use Only


Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-10
Biotin-Conjugate anti-human IL-10 monoclonal antibody
Streptavidin-HRP
Human IL-10 Standard lyophilized, 400pg/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501125351 Life Technologies
LSBMS2152CE
96 tests Each for $605.88
501125352 Life Technologies
LSBMS2152TENCE
10 x 96 tests Each for $3,945.48
Specifications

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Description & Specifications

Specifications

ELISA
TGIF, MCSF III, CSIF, B TCGF
Biotin
1.0pg/mL
50μL
IL-10
Cell culture supernatant, serum, plasma (citrate, heparin)
Human