Please login to your online account to display your discounted pricing

Human IL-10 DuoSet, R&D Systems

$317.73 - $828.14

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DY217B

 
r & d systems
RDSDY217B
15 Plates Each for $828.14

DY217B05

 
r & d systems
RDSDY217B05
5 Plates Each for $317.73
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit
Species Human

Assay Type: ELISA Development Kit
Sensitivity: 0pg/mL
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.