Please login to your online account to display your discounted pricing

Human IL-1 beta Quantikine HS ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSHSLB00C

 View more versions of this product

Catalog No. HSLB00C

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3553
Product Type Solid Phase Sandwich ELISA
Packaging Quantity 1 Plate
Quantity 1 Kit
Sensitivity 0.14pg/mL
Sample Type Serum, EDTA Plasma, Heparin Plasma
Sample Volume Serum (150μL), EDTA Plasma (150μL), Heparin Plasma (150μL)
Species Human
Specificity Natural and recombinant human IL-1 beta
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules