missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human IL-1 beta Quantikine HS ELISA Kit, R&D Systems

$780.83 - $3,747.74

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3553
Sample Type Serum, EDTA Plasma, Heparin Plasma
Sample Volume Serum (150μL), EDTA Plasma (150μL), Heparin Plasma (150μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
HSLB00C R & D Systems
RDSHSLB00C
1 Plate Each for $780.83
SSLB00C R & D Systems
RDSSSLB00C
6 Plates Each for $3,747.74
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Serum (150μL), EDTA Plasma (150μL), Heparin Plasma (150μL)
Sensitivity: 0.14pg/mL
Specificity: Natural and recombinant human IL-1 beta
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
3553
Serum (150μL), EDTA Plasma (150μL), Heparin Plasma (150μL)
Human
Solid Phase Sandwich ELISA
Serum, EDTA Plasma, Heparin Plasma
0.14pg/mL
Natural and recombinant human IL-1 beta