Please login to your online account to display your discounted pricing

Human IL-1 beta Quantikine HS ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSSSLB00C

 View more versions of this product

Catalog No. SSLB00C

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3553
Packaging Quantity 6 Plates
Quantity 1/Pk.
Sensitivity 0.14pg/mL
Sample Type Serum, EDTA Plasma, Heparin Plasma
Sample Volume Serum (150μL), EDTA Plasma (150μL), Heparin Plasma (150μL)
Species Human
Specificity Natural and recombinant human IL-1 beta
Sufficient For Sufficient materials to run ELISAs on at least six 96-well plates
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA