missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interleukin 1 beta Recombinant, Thermo Scientific™

Interleukin 1 beta Recombinant protein SIL1B from Thermo Scientific™ specifically detects Interleukin 1 beta in Not Applicable samples, and is validated for Control, ELISA and WB

$170.10 - $2,436.00

Specifications

Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Target Natural and recombinant human interleukin-1 beta (IL-1ß)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Sufficient For Price Quantity  
Catalog Number Mfr. No. Description Sufficient For Price Quantity  
ENEH2IL1B Thermo Scientific™
PIEH2IL1B
Human IL-1 beta ELISA Kit 96 microplate wells Each for $651.05
ENEH2IL1B2 Thermo Scientific™
EH2IL1B2
Human IL-1 beta ELISA Kit 2 x 96 microplate wells Each for $1,145.50
ENEH2IL1B5 Thermo Scientific™
PIEH2IL1B5
Human IL-1 beta ELISA Kit 5 x 96 microplate wells Pack of 5 Each for $2,436.00
ENESS0008 Thermo Scientific™
ESS0008
Human IL-1 beta ELISA Reagent Kit 5 x 96 microplate wells Each for $651.05
ENSIL1B Thermo Scientific™
SIL1B
Human IL-1 beta Standard for ELISA ELISA standard curve with replicates Each for $170.10
Specifications

Description

Interleukin 1 beta Recombinant protein SIL1B from Thermo Scientific™ specifically detects Interleukin 1 beta in Not Applicable samples, and is validated for Control, ELISA and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 1 Vial purified Protein (1 ng/ml) in 4% BSA and no preservative;

PA5-32681 detects C14orf1 in Human samples. PA5-32681 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-32681 The immunogen is Synthetic 15 amino acid peptide from C-Terminus of human C14orf1..
Description & Specifications

Specifications

Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Natural and recombinant human interleukin-1 beta (IL-1ß)
Absorbance at 450nm minus absorbance at 550nm
Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)