Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interleukin 1 beta Recombinant, Thermo Scientific™


Interleukin 1 beta Recombinant protein SIL1B from Thermo Scientific™ specifically detects Interleukin 1 beta in Not Applicable samples, and is validated for Control, ELISA and WB

Manufacturer: thermo scientific™  SIL1B

 View more versions of this product

Catalog No. ENSIL1B

  • / Each
 

Description & Specifications

Specifications

Precision Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Sensitivity ≤ 1 pg/mL
Formulation 1 Vial purified Protein (1 ng/ml) in 4% BSA and no preservative;
Quantity 1Vial
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Assay Range 10.2 to 400 pg/mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Sample Size 50 µL
Target Natural and recombinant human interleukin-1 beta (IL-1ß)
Description Human IL-1 beta Standard for ELISA
Specificity SIL1B
Sufficient For ELISA standard curve with replicates
Total Assay Time 4 hrs

Interleukin 1 beta Recombinant protein SIL1B from Thermo Scientific™ specifically detects Interleukin 1 beta in Not Applicable samples, and is validated for Control, ELISA and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 1 Vial purified Protein (1 ng/ml) in 4% BSA and no preservative;

PA5-32681 detects C14orf1 in Human samples. PA5-32681 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-32681 The immunogen is Synthetic 15 amino acid peptide from C-Terminus of human C14orf1..