Please login to your online account to display your discounted pricing

Human IL-1 alpha/IL-1F1 Quantikine ELISA Kit, R&D Systems

$624.40 - $2,985.15

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3552
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Sample Volume Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DLA50

 
r & d systems
RDSDLA50
1 Plate Each for $624.40

SLA50

 
r & d systems
RDSSLA50
6 Plates Each for $2,985.15
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3552
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Sample Volume Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity 1pg/mL
Species Human
Specificity Natural and recombinant human IL-1 alpha

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity: 1pg/mL
Specificity: Natural and recombinant human IL-1 alpha
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.