missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human IL-1 alpha/IL-1F1 DuoSet, R&D Systems

$324.58 - $846.01

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit
Gene ID (Entrez) 3552
Species Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
DY200 R & D Systems
RDSDY200
15 Plates Each for $846.01
DY20005 R & D Systems
RDSDY20005
5 Plates Each for $324.58
Specifications

Description

Assay Type: ELISA Development Kit
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit
3552
Human