Please login to your online account to display your discounted pricing

eBiosciencetrade; Human IgM Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IgM.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL (1:10,000 prediluted)
Synonym Immunoglobulin M
Target IgM
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgM

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125325

 
ebioscience
LSBMS2098CE
96 tests Each for $665.55

501125326

 
ebioscience
LSBMS2098FIVECE
5 x 96 tests Each for $2,449.05

501125328

 
ebioscience
LSBMS2098TENCE
10 x 96 tests Each for $4,334.05

501125329

 
ebioscience
LSBMS2098TWOCE
2 x 96 tests Each for $984.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL (1:10,000 prediluted)
Synonym Immunoglobulin M
Target IgM
Conjugate HRP
Sample Type Serum, plasma
Sensitivity 1ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.