Invitrogen™ eBioscience™ Human IgG4 Platinum ELISA Kit
For the quantitative detection of human IgG4.
Manufacturer:  Invitrogen™ LSBMS2095TEN
 View more versions of this product
Catalog No. 501125314
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human total IgG
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
HRP
10 x 96 tests
Serum, plasma
Human
Immunoglobulin G4
ELISA
0.1ng/mL
20μL (1:1,000 prediluted)
IgG4