missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ eBioscience™ Human IgG3 Platinum ELISA Kit
For the quantitative detection of human IgG3.
Manufacturer: Invitrogen™ LSBMS2094FIVE
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgG3
Description
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.Specifications
4.7-300 ng/mL | |
ELISA | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
5 x 96 tests | |
0.21 ng/mL | |
25μL (prediluted 1:10 000) | |
Human | |
1 hr 20 min | |
Human | |
3% | |
HRP |
HRP | |
Colorimetric Microplate Reader | |
ELISA | |
RUO | |
Plasma, Serum | |
Plasma (10μL), Serum (10μL) | |
2-8°C | |
1 hr 30 min | |
Immunoglobulin G2 | |
5.6% |
For Research Use Only