Invitrogen™ eBioscience™ Human IgG2 Platinum ELISA Kit
For the quantitative detection of human IgG2.
Manufacturer:  Invitrogen™ LSBMS2093FIVE
 View more versions of this product
Catalog No. 501125305
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgG2
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
HRP
5 x 96 tests
Serum, plasma
Human
Immunoglobulin G2
ELISA
0.25ng/mL
20μL (prediluted 1:100 000)
IgG2