missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human IGFBP-3 Quantikine ELISA Kit, R&D Systems

A 4.5 hour solid-phase ELISA designed to measure human IGFBP-3 in cell culture supernates, serum, and plasma. It contains NS0-expressed recombinant human IGFBP-3.

$624.40 - $2,985.15

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3486
Sample Volume Cell Culture Supernates (100μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Sensitivity 0.14ng/mL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
DGB300 R & D Systems
RDSDGB300
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $624.40
SGB300 R & D Systems
RDSSGB300
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $2,985.15
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Sensitivity: 0.14ng/mL
Specificity: Natural and recombinant human IGFBP-3
Cross Reactivity: Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules
3486
0.14ng/mL
Solid Phase Sandwich ELISA
Cell Culture Supernates (100μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Human