Please login to your online account to display your discounted pricing

Human IGF-I Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSDG100

 View more versions of this product

Catalog No. DG100

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 3479
Product Type Solid Phase Sandwich ELISA
Packaging Quantity 1 Plate
Quantity 1 Kit
Sensitivity 0.056ng/mL
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
Sample Volume Cell Culture Supernates (50μL), Serum (20μL), EDTA Plasma (20μL), Heparin Plasma (20μL)
Species Human
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate