missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human IGF-I Quantikine ELISA Kit, R&D Systems

$624.40 - $2,985.15

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 3479
Sample Volume Cell Culture Supernates (50μL), Serum (20μL), EDTA Plasma (20μL), Heparin Plasma (20μL)
Sensitivity 0.056ng/mL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
DG100 R & D Systems
RDSDG100
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $624.40
SG100 R & D Systems
RDSSG100
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $2,985.15
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (20μL), EDTA Plasma (20μL), Heparin Plasma (20μL)
Sensitivity: 0.056ng/mL
Specificity: Natural and recombinant human IGF-I
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
3479
0.056ng/mL
Solid Phase Sandwich ELISA
Cell Culture Supernates (50μL), Serum (20μL), EDTA Plasma (20μL), Heparin Plasma (20μL)
Human